Skip to Content
MilliporeSigma

BDPH9BCCED3D

Bide Pharmatech

tert-Butyl (4-oxo-4-phenylbutyl)carbamate

Synonym(s):

tert-butyl 4-oxo-4-phenylbutylcarbamate

Select a Size

Change View

About This Item

Linear Formula:
C15H21O3N1
CAS Number:
Molecular Weight:
263.34
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H21NO3/c1-15(2,3)19-14(18)16-11-7-10-13(17)12-8-5-4-6-9-12/h4-6,8-9H,7,10-11H2,1-3H3,(H,16,18)

Inchi Key

BAEFUAYRWLKGRC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Country of Origin

CN

Physical Form

White to off-white powder or crystals

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library