Skip to Content
MilliporeSigma

BDPH9BCCF87F

Bide Pharmatech

4-Phenylbutanoic acid

Synonym(s):

4-phenylbutanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H12O2
CAS Number:
Molecular Weight:
164.2
MDL number:


Inchi Code

1S/C10H12O2/c11-10(12)8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2,(H,11,12)

Inchi Key

OBKXEAXTFZPCHS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder

Melting Point

49-51C



Still not finding the right product?

Explore all of our products under 4-Phenylbutanoic acid


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library