Skip to Content

BDPH9BCCFEBF

Bide Pharmatech

4-(Trifluoromethyl)thiazol-2-amine

Synonym(s):

4-(trifluoromethyl)-1,3-thiazol-2-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H3N2S1F3
CAS Number:
Molecular Weight:
168.14
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Storage temp.

Keep in dark place, inert atmosphere, 2-8°C

Inchi Code

1S/C4H3F3N2S/c5-4(6,7)2-1-10-3(8)9-2/h1H,(H2,8,9)

Inchi Key

OVMGTNMCYLZGLS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Country of Origin

CN

Physical Form

Form: solid



Still not finding the right product?

Explore all of our products under 4-(Trifluoromethyl)thiazol-2-amine


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library