Skip to Content

BDPH9BCD19B2

Bide Pharmatech

2-Amino-5-chloro-3-fluorophenol

Synonym(s):

2-amino-5-chloro-3-fluorophenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5O1N1Cl1F1
CAS Number:
Molecular Weight:
161.56
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H5ClFNO/c7-3-1-4(8)6(9)5(10)2-3/h1-2,10H,9H2

Inchi Key

VHVCSCFCFVSSFP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2-Amino-5-chloro-3-fluorophenol


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library