Inchi Code
1S/C10H9NO2/c1-8(6-11)13-10-4-2-9(7-12)3-5-10/h2-5,7-8H,1H3
Inchi Key
QNECGSPKRWDDLX-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Colorless to Yellow Liquid or Semi-Solid or solid
Still not finding the right product?
Explore all of our products under 2-(4-Formylphenoxy)propanenitrile
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
