Inchi Code
1S/C12H13ClN2/c1-12(2,3)11-14-9-7-5-4-6-8(9)10(13)15-11/h4-7H,1-3H3
Inchi Key
RRKKSASKQFKRHK-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
White to yellow powder to crystal
Still not finding the right product?
Explore all of our products under 2-(tert-Butyl)-4-chloroquinazoline
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
