Skip to Content
MilliporeSigma

BDPH9BCD4E61

Bide Pharmatech

6-Chloro-4-iodo-1H-indazole

Synonym(s):

6-chloro-4-iodo-1H-indazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H4N2Cl1I1
Molecular Weight:
278.48
MDL number:


Inchi Code

1S/C7H4ClIN2/c8-4-1-6(9)5-3-10-11-7(5)2-4/h1-3H,(H,10,11)

Inchi Key

CZQKFAYNSDDHKA-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library