Skip to Content
MilliporeSigma

BDPH9BCDE413

Bide Pharmatech

2-(4-Bromothiophen-2-yl)-N-(4-((6-methyl-2-(pyrrolidin-1-yl)pyrimidin-4-yl)amino)phenyl)acetamide

Synonym(s):

2-(4-bromothiophen-2-yl)-N-(4-((6-methyl-2-(pyrrolidin-1-yl)pyrimidin-4-yl)amino)phenyl)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C21H22O1N5Br1S1
CAS Number:
Molecular Weight:
472.41
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C21H22BrN5OS/c1-14-10-19(26-21(23-14)27-8-2-3-9-27)24-16-4-6-17(7-5-16)25-20(28)12-18-11-15(22)13-29-18/h4-7,10-11,13H,2-3,8-9,12H2,1H3,(H,25,28)(H,23,24,26)

Inchi Key

ACDZBXZERLBYOW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals






Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library