Skip to Content
MilliporeSigma

BDPH9C021BE6

Bide Pharmatech

1-Bromo-4-((3-methylbut-3-en-2-yl)oxy)benzene

Synonym(s):

1-bromo-4-((3-methylbut-3-en-2-yl)oxy)benzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H13BrO/c1-8(2)9(3)13-11-6-4-10(12)5-7-11/h4-7,9H,1H2,2-3H3

Inchi Key

NEFCBANZVNKIBM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library