Skip to Content
MilliporeSigma

BDPH9C021F86

Bide Pharmatech

Benzyl (R)-2-(cyanomethyl)piperazine-1-carboxylate

Synonym(s):

benzyl (R)-2-(cyanomethyl)piperazine-1-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H17O2N3
CAS Number:
Molecular Weight:
259.31
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C14H17N3O2/c15-7-6-13-10-16-8-9-17(13)14(18)19-11-12-4-2-1-3-5-12/h1-5,13,16H,6,8-11H2/t13-/m1/s1

Inchi Key

QSKMFYCQRQLYMF-CYBMUJFWSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid or lump



Still not finding the right product?


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library