Skip to Content
MilliporeSigma

PG9H9BC7B2D4

PrecisePEG

s-TCO-PNP

Synonym(s):

((1R,8S,9r,E)-bicyclo[6.1.0]non-4-en-9-yl)methyl (4-nitrophenyl) carbonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C17H19O5N1
CAS Number:
Molecular Weight:
317.34
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C17H19NO5/c19-17(23-13-9-7-12(8-10-13)18(20)21)22-11-16-14-5-3-1-2-4-6-15(14)16/h1-2,7-10,14-16H,3-6,11H2/b2-1+/t14-,15+,16+

Inchi Key

JSYLBRUJIPTSJM-DTGUECQKSA-N

Shipping Temperature

ambient

Purity

96%

Storage temp.

-20

Country of Origin

CN

Physical Form

Solid



hstatements

Precautionary Statements



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library