Skip to Content
MilliporeSigma

FFT

Firefly tails (lanterns), desiccated

for bioluminescence

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
eCl@ss:
32150313
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

technique(s)

toxicology assay: suitable

storage temp.

−20°C

Application

Fireflies have been used in several studies of bioluminescence. In particular they may be used in assays to determine toxicity pathways because bioluminescence can quantitatively measure a wide range of biological activities.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library