Skip to Content
MilliporeSigma

TSTOP

ThermaStop™

increases PCR specificity, inhibits non-specific polymerase activity, prevents mis-priming before PCR, improves hot start PCR

Synonym(s):

Multiplex PCR, PCR Additive, hot start DNA polymerase, hot start PCR, hotstart

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
eCl@ss:
32150313
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized powder

packaging

pkg of 2500 units, pkg of 500 units, pkg of One unit of ThermaStop™ is defined as the amount required for maximum hot-start in amplification reactions containing one unit of Taq DNA polymerase in a volume of 25 μl.

technique(s)

PCR: suitable, whole genome amplification: suitable

color

opaque

shipped in

ambient

storage temp.

2-8°C

General description

ThermaStop™ is a reversible hot-start reagent that is compatible with thermostable DNA polymerases. It acts directly on the polymerase to prevent non-specific enzymatic activity below 50°C. Polymerase activity is fully restored at 60°C, but is inhibited again by ThermaStop™ by cooling the reaction. Adding ThermaStop™ to your PCR reactions improves sensitivity and total product yield.

Application

ThermaStop™ has been used as a component of the PCR mix for the human immunodeficiency virus (HIV-1) proviral genome amplification by non-multiplexed nested PCR. It has also been used for the amplification of DNA from oral swabs of HIV infected individuals.

Features and Benefits

  • Adding ThermaStop™ to your PCR reactions improves sensitivity and total product yield
  • Increases the product yield for downstream applications like genotyping, next-generation sequencing (NGS) sample preparation, Gibson assembly, diagnostics, and multiplex PCR
  • It improves polymerase specificity, enhances PCR performance and quantitative data
  • Prevents primer-dimer formation and non-specific products before PCR
  • Enables stabilization of master mixes at room temperature before PCR for up to 36 hrs
  • Rapidly inactivated during the first PCR cycle
  • Rapidly re-activated upon cooling

Preparation Note

Dissolve in Tris pH 8.3 buffer

Legal Information

ThermaStop is a trademark of ThermaGenix Inc.


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library