MilliporeSigma
ABAD directly links Abeta to mitochondrial toxicity in Alzheimer's disease.
Science (New York, N.Y.) (2004-04-17)
Joyce W Lustbader, Maurizio Cirilli, Chang Lin, Hong Wei Xu, Kazuhiro Takuma, Ning Wang, Casper Caspersen, Xi Chen, Susan Pollak, Michael Chaney, Fabrizio Trinchese, Shumin Liu, Frank Gunn-Moore, Lih-Fen Lue, Douglas G Walker, Periannan Kuppusamy, Zay L Zewier, Ottavio Arancio, David Stern, Shirley ShiDu Yan, Hao Wu
RESUMEN
Mitochondrial dysfunction is a hallmark of beta-amyloid (Abeta)-induced neuronal toxicity in Alzheimer's disease (AD). Here, we demonstrate that Abeta-binding alcohol dehydrogenase (ABAD) is a direct molecular link from Abeta to mitochondrial toxicity. Abeta interacts with ABAD in the mitochondria of AD patients and transgenic mice. The crystal structure of Abeta-bound ABAD shows substantial deformation of the active site that prevents nicotinamide adenine dinucleotide (NAD) binding. An ABAD peptide specifically inhibits ABAD-Abeta interaction and suppresses Abeta-induced apoptosis and free-radical generation in neurons. Transgenic mice overexpressing ABAD in an Abeta-rich environment manifest exaggerated neuronal oxidative stress and impaired memory. These data suggest that the ABAD-Abeta interaction may be a therapeutic target in AD.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
HSD17B10, GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution