MilliporeSigma
Paracrine effect of regulatory T cells promotes cardiomyocyte proliferation during pregnancy and after myocardial infarction.
Nature communications (2018-06-28)
Serena Zacchigna, Valentina Martinelli, Silvia Moimas, Andrea Colliva, Marco Anzini, Andrea Nordio, Alessia Costa, Michael Rehman, Simone Vodret, Cristina Pierro, Giulia Colussi, Lorena Zentilin, Maria Ines Gutierrez, Ellen Dirkx, Carlin Long, Gianfranco Sinagra, David Klatzmann, Mauro Giacca
RESUMEN
Cardiomyocyte proliferation stops at birth when the heart is no longer exposed to maternal blood and, likewise, to regulatory T cells (Tregs) that are expanded to promote maternal tolerance towards the fetus. Here, we report a role of Tregs in promoting cardiomyocyte proliferation. Treg-conditioned medium promotes cardiomyocyte proliferation, similar to the serum from pregnant animals. Proliferative cardiomyocytes are detected in the heart of pregnant mothers, and Treg depletion during pregnancy decreases both maternal and fetal cardiomyocyte proliferation. Treg depletion after myocardial infarction results in depressed cardiac function, massive inflammation, and scarce collagen deposition. In contrast, Treg injection reduces infarct size, preserves contractility, and increases the number of proliferating cardiomyocytes. The overexpression of six factors secreted by Tregs (Cst7, Tnfsf11, Il33, Fgl2, Matn2, and Igf2) reproduces the therapeutic effect. In conclusion, Tregs promote fetal and maternal cardiomyocyte proliferation in a paracrine manner and improve the outcome of myocardial infarction.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anticuerpo anti-fosfo-histona H3 (Ser10), marcador de mitosis, Upstate®, from rabbit
Sigma-Aldrich
Anti-PCM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2