MilliporeSigma
  • Hydrated lanthanum oxide-modified diatomite as highly efficient adsorbent for low-concentration phosphate removal from secondary effluents.
Hydrated lanthanum oxide-modified diatomite as highly efficient adsorbent for low-concentration phosphate removal from secondary effluents.
Journal of environmental management (2018-10-28)
You Wu, Xiaoming Li, Qi Yang, Dongbo Wang, Qiuxiang Xu, Fubing Yao, Fei Chen, Ziletao Tao, Xiaoding Huang
RESUMEN
The requirement to the phosphorus (P) emission from wastewater treatment plants (WWTPs) is becoming increasingly strict, which makes an advanced treatment for the low-concentration phosphate removal from secondary effluents indispensable. In present work, hydrated lanthanum (La) oxide-modified diatomite composites (La-diatomite) were fabricated by a facile method and employed as the highly efficient adsorbent for the low-concentration phosphate removal from simulating secondary effluents. Comparative experiments indicated that the La-diatomite treated by 0.1 mol/L LaCl3 exhibited the highest La availability (P/La molar ratio of 2.30) and performed good selectivity to phosphate adsorption even with the coexistence of competing anions and humic acid. The maximum P adsorption capacity reached to 58.7 mg P/g and the 96% P was removed quickly within 30 min at initial phosphate concentration 2 mg P/L. Insignificant La leaching was observed during the process due to the La stabilization by macroporous diatomite. Eight cycles of adsorption-desorption experiments revealed that the excellent repeated use property of La-diatomite. At the column test, La-diatomite showed superior treatment capacities of 3455 kg water/kg La-diatomite for simulated secondary effluents. The La-diatomite maintained high and stable adsorption effectiveness in wide pH range, which should be attributed to the synergistic effect of electrostatic interactions, ligand exchange and Lewis acid-based interaction. This work might provide a candidate for low-concentration phosphate removal from secondary effluent to alleviate the eutrophication.