MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Nogo-A-targeting antibody promotes visual recovery and inhibits neuroinflammation after retinal injury.
Cell death & disease (2020-02-08)
Julius Baya Mdzomba, Sandrine Joly, Léa Rodriguez, Ali Dirani, Patricia Lassiaz, Francine Behar-Cohen, Vincent Pernet
RESUMEN
N-Methyl-D-aspartate (NMDA)-induced neuronal cell death is involved in a large spectrum of diseases affecting the brain and the retina such as Alzheimer's disease and diabetic retinopathy. Associated neurological impairments may result from the inhibition of neuronal plasticity by Nogo-A. The objective of the current study was to determine the contribution of Nogo-A to NMDA excitotoxicity in the mouse retina. We observed that Nogo-A is upregulated in the mouse vitreous during NMDA-induced inflammation. Intraocular injection of a function-blocking antibody specific to Nogo-A (11C7) was carried out 2 days after NMDA-induced injury. This treatment significantly enhanced visual function recovery in injured animals. Strikingly, the expression of potent pro-inflammatory molecules was downregulated by 11C7, among which TNFα was the most durably decreased cytokine in microglia/macrophages. Additional analyses suggest that TNFα downregulation may stem from cofilin inactivation in microglia/macrophages. 11C7 also limited gliosis presumably via P.Stat3 downregulation. Diabetic retinopathy was associated with increased levels of Nogo-A in the eyes of donors. In summary, our results reveal that Nogo-A-targeting antibody can stimulate visual recovery after retinal injury and that Nogo-A is a potent modulator of excitotoxicity-induced neuroinflammation. These data may be used to design treatments against inflammatory eye diseases.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anticuerpo anti-colina acetiltransferasa, Chemicon®, from goat
Sigma-Aldrich
Anticuerpo anti-Olig-2, Chemicon®, from rabbit
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
Anticuerpo anti-glutamina sintetasa, clon GS-6, clone GS-6, Chemicon®, from mouse