MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Comprehensive and quantitative profiling of B vitamins and related compounds in the mammalian liver.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2019-12-11)
Juan Xu, Constance E Clare, Amey H Brassington, Kevin D Sinclair, David A Barrett
RESUMEN
A method for the simultaneous quantification of B vitamins and related amines in one-carbon (1C) metabolism would benefit the study of diet and genetic/epigenetic regulation of mammalian development and health. We present a validated method for the simultaneous quantitative analysis of 13 B vitamers and four related 1C-pathway amine intermediates in liver using hydrophilic interaction chromatography (HILIC) coupled to electrospray ionization tandem mass spectrometry. Frozen sheep liver samples (50 mg) were homogenized in cold 50% acetonitrile containing 1% acetic acid with the addition of two isotope labelled internal standards. Hot acid hydrolysis was applied to release the protein-bound forms. The separation of 17 analytes was achieved using a pHILIC column with a total run time of 13 min. Detection was achieved in electrospray positive ionisation mode. Limits of detection for the majority of analytes were within the range of 0.4-3.2 pmol/g. The method was applied to 266 sheep liver samples and revealed that adenosylcobalamin, methylcobalamin, pyridoxic acid, flavin adenine dinucleotide and thiamine were the major forms of the B vitamers present with pyridoxal 5'-phosphate and thiamine pyrophosphate being detected at lower concentrations. Trimethylglycine and methylglycine were the predominant 1C-related amines measured. As anticipated, the B vitamin status of individuals varied considerably, reflecting dietary and genetic variation in our chosen outbred model species. This method offers a simple sample extraction procedure and provides comprehensive coverage of B vitamins coupled with good sensitivity and reliability.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Pyridoxine, ≥98%
Sigma-Aldrich
Coenzyme B12, ≥97.0%
Sigma-Aldrich
4-Pyridoxic acid, ≥98%
Sigma-Aldrich
Trimethylgallium, packaged for use in deposition systems