MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • PKR inhibitors suppress endoplasmic reticulum stress and subdue glucolipotoxicity-mediated impairment of insulin secretion in pancreatic beta cells.
PKR inhibitors suppress endoplasmic reticulum stress and subdue glucolipotoxicity-mediated impairment of insulin secretion in pancreatic beta cells.
Turkish journal of biology = Turk biyoloji dergisi (2020-04-08)
Abdullah Yalçin, Gülçin Şarkici, Umut Kerem Kolaç
RESUMEN
Type 2 diabetes mellitus is characterized by insulin resistance and hypersecretion of insulin from the pancreas to compensate for decreased insulin sensitivity in the peripheral tissues. In later stages of the disease insulin-secreting beta cell degeneration commences and patients require insulin replacement therapy in order to accomplish proper regulation of their blood glucose. Endoplasmic reticulum (ER) stress in the beta cells is one of the factors contributing to this detrimental effect. Protein kinase R (PKR) is a cellular stress kinase activated by ER stress and contributing to degeneration of pancreatic islets. In order to determine whether inhibition of PKR activation by specific small molecule inhibitors of PKR ameliorates pancreatic insulin secretion capacity, we treated beta cells with two imidazole/oxindole-derived inhibitors of PKR kinase, imoxin (C16) and 2-aminopurine (2-AP), in the presence of ER stress. Our results demonstrate that PKR inhibition suppresses tunicamycin-mediated ER stress without altering the insulin production capacity of the cells. Palmitic acid-mediated suppression of insulin secretion, however, was subdued significantly by PKR inhibitor treatment through an ER stress-related mechanism. We suggest that PKR inhibitor treatment may be used to increase the insulin secretion capacity of the pancreas in later stages of diabetes.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Penicilina- Estreptomicina, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamina solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
Sodium palmitate, ≥98.5%
Sigma-Aldrich
2-Aminopurine, ≥99%
Sigma-Aldrich
Imidazolo-oxindole PKR inhibitor C16, ≥98% (HPLC)
Sigma-Aldrich
Human PLG (Plasminogen) ELISA Kit, for cell culture supernatants, plasma, and serum samples
Sigma-Aldrich
Bovine INS / Insulin ELISA Kit