MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Effect of ensiling duration on the fate of deoxynivalenol, zearalenone and their derivatives in maize silage.
Mycotoxin research (2019-11-11)
Tolke Jensen, Marthe De Boevre, Sarah De Saeger, Nils Preußke, Frank D Sönnichsen, Ewald Kramer, Holger Klink, Joseph-Alexander Verreet, Tim Birr
RESUMEN
Fusarium mycotoxins and their derivatives are frequently detected in freshly harvested forage maize. This study assessed the time course effects during ensiling of forage maize on the fate of Fusarium mycotoxins, using laboratory-scale silos and artificially contaminated raw material. A multi-mycotoxin liquid chromatography-high-resolution mass spectrometry (LC-HRMS) method was used to determine the levels of deoxynivalenol (DON), zearalenone (ZEN) and their derivatives DON-3-glucoside, 3-acetyl-DON, 15-acetyl-DON, deepoxy-DON, α-zearalenol and β-zearalenol. A significant increase of DON was observed during ensiling, whereas the levels of DON-3-glucoside and its acetylated forms proportionally decreased. In contrast, levels of ZEN, α-zearalenol and β-zearalenol were not affected by the ensiling process. Based on these findings, ensiling is not a practical method for reducing the total amount of Fusarium mycotoxins present at harvest.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
6-Diazo-5-oxo-L-norleucine, powder
Supelco
Deepoxy-deoxynivalenol solution, ~50 μg/mL in acetonitrile, analytical standard