MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Arrays of lipid bilayers and liposomes on patterned polyelectrolyte templates.
Journal of colloid and interface science (2006-06-23)
Neeraj Kohli, Sachin Vaidya, Robert Y Ofoli, Robert M Worden, Ilsoon Lee
RESUMEN
This paper presents novel methods to produce arrays of lipid bilayers and liposomes on patterned polyelectrolyte multilayers. We created the arrays by exposing patterns of poly(dimethyldiallylammonium chloride) (PDAC), polyethylene glycol (m-dPEG) acid, and poly(allylamine hydrochloride) (PAH) on polyelectrolyte multilayers (PEMs) to liposomes of various compositions. The resulting interfaces were characterized by total internal reflection fluorescence microscopy (TIRFM), fluorescence recovery after pattern photobleaching (FRAPP), quartz crystal microbalance (QCM), and fluorescence microscopy. Liposomes composed of 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC) and 1,2-dioleoyl-sn-glycero-3-phosphate (monosodium salt) (DOPA) were found to preferentially adsorb on PDAC and PAH surfaces. On the other hand, liposome adsorption on sulfonated poly(styrene) (SPS) surfaces was minimal, due to electrostatic repulsion between the negatively charged liposomes and the SPS-coated surface. Surfaces coated with m-dPEG acid were also found to resist liposome adsorption. We exploited these results to create arrays of lipid bilayers by exposing PDAC, PAH and m-dPEG patterned substrates to DOPA/DOPC vesicles of various compositions. The patterned substrates were created by stamping PDAC (or PAH) on SPS-topped multilayers, and m-dPEG acid on PDAC-topped multilayers, respectively. This technique can be used to produce functional biomimetic interfaces for potential applications in biosensors and biocatalysis, for creating arrays that could be used for high-throughput screening of compounds that interact with cell membranes, and for probing, and possibly controlling, interactions between living cells and synthetic membranes.