MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Alkaloids from Daphniphyllum oldhami.
Journal of natural products (2008-02-22)
Shu-Zhen Mu, Jun-Song Wang, Xiao-Sheng Yang, Hong-Ping He, Chun-Shun Li, Ying-Tong Di, Ye Wang, Yu Zhang, Xin Fang, Lie-Jun Huang, Xiao-Jiang Hao
RESUMEN
Four new Daphniphyllum alkaloids, daphnioldhanines H-K ( 1- 4), along with 34 known alkaloids, were isolated from Daphniphyllum oldhami. The known alkaloid dehydrodaphnigraciline ( 5) is now reported as a natural product. Their structures were elucidated by spectroscopic methods, especially 2D NMR techniques. The effects against platelet aggregation of compounds 1, 3, and 5 were evaluated, and 3 showed stronger activity against platelet aggregation induced by PAF. This is the first report of quinolizidine alkaloids from the genus Daphniphyllum.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Acetylsalicylic acid, analytical standard