MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Prevalence and diversity of enteric pathogens among cholera treatment centre patients with acute diarrhea in Uvira, Democratic Republic of Congo.
Prevalence and diversity of enteric pathogens among cholera treatment centre patients with acute diarrhea in Uvira, Democratic Republic of Congo.
BMC infectious diseases (2020-10-11)
Camille Williams, Oliver Cumming, Lynn Grignard, Baron Bashige Rumedeka, Jaime Mufitini Saidi, Daniel Grint, Chris Drakeley, Aurelie Jeandron
RESUMEN
Cholera remains a major global health challenge. Uvira, in the Democratic Republic of the Congo (DRC), has had endemic cholera since the 1970's and has been implicated as a possible point of origin for national outbreaks. A previous study among this population, reported a case confirmation rate of 40% by rapid diagnostic test (RDT) among patients at the Uvira Cholera Treatment Centre (CTC). This study considers the prevalence and diversity of 15 enteric pathogens in suspected cholera cases seeking treatment at the Uvira CTC. We used the Luminex xTAG® multiplex PCR to test for 15 enteric pathogens, including toxigenic strains of V. cholerae in rectal swabs preserved on Whatman FTA Elute cards. Results were interpreted on MAGPIX® and analyzed on the xTAG® Data Analysis Software. Prevalence of enteric pathogens were calculated and pathogen diversity was modelled with a Poisson regression. Among 269 enrolled CTC patients, PCR detected the presence of toxigenic Vibrio cholerae in 38% (103/269) of the patients, which were considered to be cholera cases. These strains were detected as the sole pathogen in 36% (37/103) of these cases. Almost half (45%) of all study participants carried multiple enteric pathogens (two or more). Enterotoxigenic Escherichia coli (36%) and Cryptosporidium (28%) were the other most common pathogens identified amongst all participants. No pathogen was detected in 16.4% of study participants. Mean number of pathogens was highest amongst boys and girls aged 1-15 years and lowest in women aged 16-81 years. Ninety-three percent of toxigenic V. cholerae strains detected by PCR were found in patients having tested positive for V. cholerae O1 by RDT. Our study supports previous results from DRC and other cholera endemic areas in sub-Sahara Africa with less than half of CTC admissions positive for cholera by PCR. More research is required to determine the causes of severe acute diarrhea in these low-resource, endemic areas to optimize treatment measures. This study is part of the impact evaluation study entitled: "Impact Evaluation of Urban Water Supply Improvements on Cholera and Other Diarrheal Diseases in Uvira, Democratic Republic of Congo" registered on 10 October 2016 at clinicaltrials.gov Identification number: NCT02928341 .
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)