MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Synthesis and biological evaluation of arylidene analogues of Meldrum's acid as a new class of antimalarial and antioxidant agents.
Synthesis and biological evaluation of arylidene analogues of Meldrum's acid as a new class of antimalarial and antioxidant agents.
Bioorganic & medicinal chemistry (2010-07-14)
Harmeet S Sandhu, Sameer Sapra, Mukesh Gupta, Kunal Nepali, Raju Gautam, Sunil Yadav, Raj Kumar, Sanjay M Jachak, Manoj Chugh, Manish K Gupta, Om P Suri, K L Dhar
RESUMEN
A series of arylidene analogues of Meldrum's acid were synthesized and evaluated for in vitro antimalarial and antioxidant activities for the first time. The influence of various physico-chemical parameters such as dielectric constant (epsilon), donor number (DN), acceptor number (AN), hydrogen bond donor (HBD), hydrogen bond acceptor (HBA), and solubilizing power of the solvents on Meldrum's acid anion generation and thus on promoting the Knoevenagel condensation of Meldrum's acid with aryl aldehydes has been discussed. Five compounds 9l, 9m, 9n, 9r, and 9s were found to be most active against Plasmodiumfalciparum with IC(50) values in the range of 9.68-16.11 microM. Compound 9l exhibited the most potent antimalarial activity (IC(50) 9.68 microM). The compounds were also found to possess antioxidant activity when tested against DPPH and ABTS free radicals.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido L-ascórbico, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Ácido L-ascórbico, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Ácido L-ascórbico, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Ácido L-ascórbico, 99%
Sigma-Aldrich
Ácido L-ascórbico, reagent grade, crystalline
Sigma-Aldrich
Ácido L-ascórbico, ACS reagent, ≥99%
Supelco
Ácido L-ascórbico, analytical standard
Sigma-Aldrich
Ácido L-ascórbico, meets USP testing specifications
Sigma-Aldrich
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
Sigma-Aldrich
Ácido L-ascórbico, reagent grade
Sigma-Aldrich
Ácido L-ascórbico, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Ácido L-ascórbico, FCC, FG
Sigma-Aldrich
Ácido L-ascórbico, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Ácido L-ascórbico, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Ácido L-ascórbico, tested according to Ph. Eur.