MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Synthesis and antioxygenic activities of seabuckthorn flavone-3-ols and analogs.
Bioorganic & medicinal chemistry letters (2011-08-09)
N Pandurangan, Chinchu Bose, A Banerji
RESUMEN
A practical synthesis of polyhydroxy- and regiospecifically methylated flavone-3-ols which are components of commercial 'seabuckthorn flavone' has been achieved by modified Algar-Flynn-Oyamada method. Antioxidant activities of seabuckthorn extracts, isolated products and a number of flavone-3-ols have been determined. Structure-activity relationships have been discussed. Amongst the compounds tested, gallic acid, which is also present in seabuckthorn, was found to be the most effective antioxidant and radioprotectant.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido gálico, 97.5-102.5% (titration)
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
Sigma-Aldrich
Gallic acid monohydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Gallic acid monohydrate, ≥99% (HPLC)
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Isorhamnetin, ≥95.0% (HPLC)