MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Synthesis and antioxidant activity of long chain alkyl hydroxycinnamates.
European journal of medicinal chemistry (2011-01-11)
Jose C J M D S Menezes, Shrivallabh P Kamat, Jose A S Cavaleiro, Alexandra Gaspar, Jorge Garrido, Fernanda Borges
RESUMEN
Long chain alkyl hydroxycinnamates (8-21) were synthesized from the corresponding half esters of malonic acid (5-7) and benzaldehyde derivatives by Knoevenagel condensation. The total antioxidant capacity of these hydroxycinnamyl esters was evaluated using DPPH and ABTS assays. The observed antioxidant activity was highest for esters of caffeic acid followed by sinapic esters and ferulic esters. The parameters for drug-likeness of these hydroxycinnamyl esters were also evaluated according to the Lipinski's 'rule-of-five'. All the ester derivatives were found to violate one of the Lipinski's parameters (cLogP>5), even though they have been found to be soluble in protic solvents. The predictive topological polar surface area (TPSA) data allow concluding that they could have a good capacity for penetrating cell membranes. Therefore, one can propose these novel lipophilic compounds as potential antioxidants for tackling oxidative processes.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Ferulic acid, 99%
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Ferulic acid, ≥99%
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Supelco
Ferulic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Supelco
Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland