MilliporeSigma
PUPIL enables mapping and stamping of transient electrical connectivity in developing nervous systems.
Cell reports (2021-10-24)
Shu Xie, Haixiang Li, Fenyong Yao, Jiechang Huang, Xiaomei Yang, Xin Chen, Qiang Liu, Min Zhuang, Shuijin He
RESUMEN
Currently, many genetic methods are available for mapping chemical connectivity, but analogous methods for electrical synapses are lacking. Here, we present pupylation-based interaction labeling (PUPIL), a genetically encoded system for noninvasively mapping and stamping transient electrical synapses in the mouse brain. Upon fusion of connexin 26 (CX26) with the ligase PafA, pupylation yields tag puncta following conjugation of its substrate, a biotin- or fluorescent-protein-tagged PupE, to the neighboring proteins of electrical synapses containing CX26-PafA. Tag puncta are validated to correlate well with functional electrical synapses in immature neurons. Furthermore, puncta are retained in mature neurons when electrical synapses mostly disappear-suggesting successful stamping. We use PUPIL to uncover spatial subcellular localizations of electrical synapses and approach their physiological functions during development. Thus, PUPIL is a powerful tool for probing electrical connectivity patterns in complex nervous systems and has great potential for transient receptors and ion channels as well.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Doxiciclina hyclate
Sigma-Aldrich
Anti-Connexin 35/36 Antibody, clone 9D7.2, clone 9D7.2, Chemicon®, from mouse