MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • 2-Hydroxypropyl-β-cyclodextrin is the active component in a triple combination formulation for treatment of Niemann-Pick C1 disease.
2-Hydroxypropyl-β-cyclodextrin is the active component in a triple combination formulation for treatment of Niemann-Pick C1 disease.
Biochimica et biophysica acta. Molecular and cell biology of lipids (2019-05-06)
Jessica Davidson, Elizabeth Molitor, Samantha Moores, Sarah E Gale, Kanagaraj Subramanian, Xuntian Jiang, Rohini Sidhu, Pamela Kell, Jesse Zhang, Hideji Fujiwara, Cristin Davidson, Paul Helquist, Bruce J Melancon, Michael Grigalunas, Gang Liu, Farbod Salahi, Olaf Wiest, Xin Xu, Forbes D Porter, Nina H Pipalia, Dana L Cruz, Edward B Holson, Jean E Schaffer, Steven U Walkley, Frederick R Maxfield, Daniel S Ory
RESUMEN
Niemann-Pick type C1 (NPC1) disease is a fatal neurovisceral disease for which there are no FDA approved treatments, though cyclodextrin (HPβCD) slows disease progression in preclinical models and in an early phase clinical trial. Our goal was to evaluate the mechanism of action of a previously described combination-therapy, Triple Combination Formulation (TCF) - comprised of the histone deacetylase inhibitor (HDACi) vorinostat/HPβCD/PEG - shown to prolong survival in Npc1 mice. In these studies, TCF's benefit was attributed to enhanced vorinostat pharmacokinetics (PK). Here, we show that TCF reduced lipid storage, extended lifespan, and preserved neurological function in Npc1 mice. Unexpectedly, substitution of an inactive analog for vorinostat in TCF revealed similar efficacy. We demonstrate that the efficacy of TCF was attributable to enhanced HPβCD PK and independent of NPC1 protein expression. We conclude that although HDACi effectively reduce cholesterol storage in NPC1-deficient cells, HDACi are ineffective in vivo in Npc1 mice.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
(2-Hidroxipropil)-β-ciclodextrina, powder