MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Characterization of protein-protein interactions by isothermal titration calorimetry.
Methods in molecular biology (Clifton, N.J.) (2004-04-06)
Adrian Velazquez-Campoy, Stephanie A Leavitt, Ernesto Freire
RESUMEN
Isothermal titration calorimetry (ITC) is a powerful technique to study both protein-ligand and protein-protein interactions. This methods chapter is devoted to describing protein-protein interactions, in particular, the association between two different proteins and the self-association of a protein into homodimers. ITC is the only technique that determines directly the thermodynamic parameters of a given reaction: DeltaG, DeltaH, DeltaS, and DeltaCP. Isothermal titration calorimeters have evolved over the years and one of the latest models is the VP-ITC produced by Microcal, Inc. In this chapter we will be describing the general procedure for performing an ITC experiment as well as for the specific cases of porcine pancreatic trypsin binding to soybean trypsin inhibitor and the dissociation of bovine pancreatic alpha-chymotrypsin.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Tripsina from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type I-S, essentially salt-free, lyophilized powder