MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Proliferation and osteo/odontoblastic differentiation of stem cells from dental apical papilla in mineralization-inducing medium containing additional KH(2)PO(4).
Proliferation and osteo/odontoblastic differentiation of stem cells from dental apical papilla in mineralization-inducing medium containing additional KH(2)PO(4).
Cell proliferation (2013-03-21)
L Wang, M Yan, Y Wang, G Lei, Y Yu, C Zhao, Z Tang, G Zhang, C Tang, J Yu, H Liao
RESUMEN
Stem cells from the dental apical papilla (SCAPs) can be induced to differentiate along both osteoblast and odontoblast lineages. However, little knowledge is available concerning their differentiation efficiency in osteogenic media containing additional KH2 PO4 . Stem cells from the dental apical papilla were isolated from apical papillae of immature third molars and treated with two kinds of mineralization-inducing media, MM1 and MM2, differing in KH2 PO4 concentration. Proliferation and osteo/odontogenic differentiation capacity of MM1/MM2-treated SCAPs were investigated and compared both in vitro and in vivo. Cell counting and flow cytometry demonstrated that MM2 containing 1.8 mm additional KH2 PO4 significantly enhanced proliferative potential of SCAPs, compared to MM1. Osteo/odontogenic capacity of SCAPs was much better in MM2 medium than in MM1, as indicated by elevated alkaline phosphatase activity, increased calcium deposition and upregulated expression of osteo/odontoblast-specific genes/proteins (for example, runt-related transcription factor 2, osterix, osteocalcin, dentin sialoprotein, and dentin sialophosphoprotein). In vivo transplantation findings proved that SCAPs in MM2 group generated more mineralized tissues, and presented higher expression of osteo/odontoblast-specific proteins (osteocalcin and dentin sialoprotein) than those in the MM1 group. Mineralization-inducing media supplemented with 1.8 mm additional KH2 PO4 significantly enhanced cell proliferation and improved differentiation capacity of SCAPs along osteo/odontogenic cell lineages, compared to counterparts lacking additional KH2 PO4 .
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Fosfato de potasio monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Fosfato de potasio dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Fosfato de potasio monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Fosfato de potasio monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Fosfato de potasio monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%
Sigma-Aldrich
Fosfato de potasio monobasic, ReagentPlus®
Sigma-Aldrich
Fosfato de potasio monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Fosfato de potasio dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Fosfato de potasio dibasic, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Fosfato de potasio dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Fosfato de potasio monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate monobasic solution, reagent grade, 1.0 M in solution
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Fosfato de potasio monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Fosfato de potasio monobasic, 99.99% trace metals basis
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Fosfato de potasio dibasic, 99.95% trace metals basis
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Potassium phosphate monobasic solution, 1000 ppm P
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host