MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
pH-dependence of complexion constants and complex mobility in capillary electrophoresis separations of dipeptide enantiomers.
Electrophoresis (2001-10-09)
S Sabbah, F Süss, G K Scriba
RESUMEN
The chiral separation of the LL- and DD-enantiomers of the dipeptides Ala-Tyr, Phe-Phe, and Asp-PheOMe has been investigated at pH 2.5 and pH 3.5 using beta-cyclodextrin (beta-CD), heptakis-(2,6-di-O-methyl)-beta-cyclodextrin, and heptakis-(2,3,6-tri-O-methyl)-beta-cyclodextrin as chiral selectors. According to electrospray mass spectrometry, heptakis-(2,6-di-O-methyl)-beta-cyclodextrin was a mixture of six isomers. Reversal of the enantiomer migration order upon increasing the buffer pH from 2.5 to 3.5 was observed for all peptides with beta-cyclodextrin, for Ala-Tyr and Phe-Phe in the presence of heptakis-(2,3,6-tri-O-methyl)-beta-cyclodextrin, and for Ala-Tyr using heptakis-(2,6-di-O-methyl)-beta-cyclodextrin. The migration behavior could be explained on the basis of the complexation constants and the mobilities of the peptide-cyclodextrin complexes. Both, the binding constants and complex mobilities decreased with increasing pH as the overall-charge of the peptides decreased. While the complexation constants primarily determined the migration order at pH 2.5, complex mobility dominated in most cases at pH 3.5.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Asp-Phe, 96%
Sigma-Aldrich
Phe-Phe
Sigma-Aldrich
Ala-Tyr
Sigma-Aldrich
Heptakis(2,3,6-tri-O-methyl)-β-cyclodextrin, ≥98.0%
Sigma-Aldrich
Heptakis(2,3,6-tri-O-methyl)-β-cyclodextrin, ≥90%