MilliporeSigma
Debromination of hexabromobenzene by its co-grinding with CaO.
Chemosphere (2002-09-12)
Qiwu Zhang, Hiroki Matsumoto, Fumio Saito, Michel Baron
RESUMEN
A hexabromobenzene (HBB, C6Br6) powder was subjected to co-grinding with excess CaO powder in air using a planetary ball mill to investigate mechanochemical debromination of the HBB sample. The reaction proceeds smoothly with an increase in both grinding time and molar ratio of CaO addition. The co-grinding enables us to debrominate HBB, forming CaBr2, C and CaCO3 besides excess CaO in the product. CaBr2 in the product tends to absorb moisture from the atmosphere to form CaBr2 x nH2O. The reaction yield is quantitatively evaluated by measuring the amount of water-soluble bromide through water washing and the amount of unreacted HBB through toluene washing. It is found that almost 100% of debromination is achieved by 6 h grinding with the molar ratio kept constant at 2:1. Radicals are detected in the ground products, possibly due to rupture in the covalent bonds of C-C and C-Br in HBB.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Hexabromobenzene, 98%