MilliporeSigma
  • Short-term exposure of Chinook salmon (Oncoryhnchus tshawytscha) to o,p-DDE or DMSO during early life-history stages causes long-term humoral immunosuppression.
Short-term exposure of Chinook salmon (Oncoryhnchus tshawytscha) to o,p-DDE or DMSO during early life-history stages causes long-term humoral immunosuppression.
Environmental health perspectives (2003-10-11)
Ruth H Milston, Martin S Fitzpatrick, Anthony T Vella, Shaun Clements, Deke Gundersen, Grant Feist, Tawni L Crippen, Joann Leong, Carl B Schreck
RESUMEN
We evaluated the effect of short-term exposures to a xenobiotic chemical during early life-history stages on the long-term immune competence of chinook salmon (Oncoryhnchus tshawytscha). Immersion of chinook salmon eggs in a nominal concentration of o,p-dichlorodiphenyldichloroethylene (o,p-DDE; 10 ppm) for 1 hr at fertilization followed by immersion in the same dose for 2 hr at hatch resulted in a significant reduction in the ability of splenic leukocytes from fish 1 year after treatment to undergo blastogenesis upon in vitro stimulation with lipopolysaccharide. We also observed that the vehicle, dimethyl sulfoxide (DMSO), caused a significant reduction in the ability of the splenic leukocytes to express surface immunoglobin M (SIgM) at this time. The concentration of o,p-DDE in a pooled sample of whole fry from this treatment was 0.53 microg/g lipid 1 month after first feeding but was undetectable in all other treatments. Mortality rate, time to hatch, fish length, and weight were unaffected by treatment with o,p-DDE. Similarly, sex ratios, gonadal development, and concentrations of plasma estradiol and 11-ketotestosterone were not affected by the treatment. In addition, we found no evidence that plasma lysozyme concentrations or the mitogenic responses of splenic leukocytes to concanavalin A or polyinosinic-polycytidylic acid were influenced by the treatment. In this experiment, a brief period of exposure to o,p-DDE or DMSO during early development was able to induce long-term effects on humoral immune competence of chinook salmon. Such immunosuppression may increase susceptibility to disease, which may in turn be critical to regulating the population.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
2,4′-DDE solution, 100 μg/mL in methanol, PESTANAL®, analytical standard