MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
New analogues of butylated hydroxytoluene as anti-inflammatory and antioxidant agents.
Bioorganic & medicinal chemistry (2006-05-13)
George N Ziakas, Eleni A Rekka, Antonios M Gavalas, Phaedra T Eleftheriou, Panos N Kourounakis
RESUMEN
Amine or amide derivatives bearing the 2,6-di-tert-butyl phenol moiety are synthesised. Almost all are antioxidants, reduce acute inflammation and inhibit COX-1 and lipoxygenase activity. The most potent anti-inflammatory, COX-1 inhibitor and antioxidant agent, with low toxicity, is 2,6-di-tert-butyl-4-thiomorpholin-4-ylmethyl-phenol.