MilliporeSigma
Serum activity of dipeptidyl peptidase IV (DPPIV; EC 3.4.14.5) in breast-fed infants with symptoms of allergy.
Peptides (2007-01-06)
Beata Jarmołowska, Krzysztof Bielikowicz, Małgorzata Iwan, Katarzyna Sidor, Elzbieta Kostyra, Maciej Kaczmarski
RESUMEN
Beta-casomorphins, opioid peptides present in mother's milk, are a good substrate for DPPIV (EC 3.4.14.5) which is a major factor limiting the half-life of biologically active peptides. Serum DPPIV activity of two groups of infants (healthy and atopic dermatitis) and contents of beta-casomorphin-5 and -7 in their mothers' milk were determined in the study. We have found correlation between those two parameters in the group of children with atopic dermatitis syndromes, while no such a correlation was found in the control group.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
β-Casomorphin Fragment 1-5 hydrochloride, ≥97% (HPLC)