MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Experimental correlation between the pKa value of sulfonphthaleins with the nature of the substituents groups.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-08-25)
Patricia Balderas-Hernández, María Teresa Ramírez-Silva, Mario Romero-Romo, Manuel Palomar-Pardavé, Gabriela Roa-Morales, Carlos Barrera-Diaz, Alberto Rojas-Hernández
RESUMEN
This work presents the results obtained from a spectrophotometry study performed on some indicators of the sulfonphtaleins like phenol red (PR), thymol blue (TB), bromothymol blue (BTB), xylenol orange (XO) and methylthymol blue (MTB). During the first stage the acidity constants of some of the indicators were determined using the data from spectrophotometry, potentiometry and with the use of the software SQUAD. These were as follows: for the equilibrium 2H+BTB<-->H(2)BTB, log beta(2)=15.069+/-0.046 and for H+BTB<-->HBTB, log beta(1)=8.311+/-0.044. For the XO and the MTB five values were calculated for each, namely, for MTB: log beta(5)=42.035, log beta(4)=38.567+/-0.058, log beta(3)=32.257+/-0.057, log beta(2)=23.785+/-0.057, and log beta(1)=12.974+/-0.045 while for XO: log beta(5)=40.120+/-0.102, log beta(4)=35.158+/-0.062, log beta(3)=29.102+/-0.053, log beta(2)=21.237+/-0.044, and log beta(1)=11.682+/-0.044. During the second stage, a study was conducted on the effect of the substituents present in the indicators to determine the effect of different functional groups on the pK(a) value corresponding to the last indicator's dissociation.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Bromothymol Blue, ACS reagent, Dye content 95 %
Sigma-Aldrich
Thymol Blue, ACS reagent
Sigma-Aldrich
Bromothymol Blue solution, pH 6.0-7.6
Supelco
Thymolphthalein, ACS reagent
Sigma-Aldrich
Thymolphthalein, ACS reagent, Dye content 95 %
Sigma-Aldrich
Thymol Blue sodium salt, ACS reagent, Dye content 95 %
Sigma-Aldrich
Bromothymol Blue, JIS special grade
Sigma-Aldrich
Bromothymol Blue solution, pH 6.0-7.6
Sigma-Aldrich
Thymol Blue, JIS special grade
Sigma-Aldrich
Thymolphthalein, JIS special grade