MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Monitoring bioreactors using principal component analysis: production of penicillin G acylase as a case study.
Bioprocess and biosystems engineering (2009-09-04)
Edson Romano Nucci, Antonio J G Cruz, Roberto C Giordano
RESUMEN
The complexity of biological processes often makes impractical the development of detailed, structured phenomenological models of the cultivation of microorganisms in bioreactors. In this context, data pre-treatment techniques are useful for bioprocess control and fault detection. Among them, principal component analysis (PCA) plays an important role. This work presents a case study of the application of this technique during real experiments, where the enzyme penicillin G acylase (PGA) was produced by Bacillus megaterium ATCC 14945. PGA hydrolyzes penicillin G to yield 6-aminopenicilanic acid (6-APA) and phenyl acetic acid. 6-APA is used to produce semi-synthetic beta-lactam antibiotics. A static PCA algorithm was implemented for on-line detection of deviations from the desired process behavior. The experiments were carried out in a 2-L bioreactor. Hotteling's T(2) was the discrimination criterion employed in this multivariable problem and the method showed a high sensibility for fault detection in all real cases that were studied.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
(+)-6-Aminopenicillanic acid, 96%