MilliporeSigma
[The influence of estazolam on human neurobehavioral function].
Fa yi xue za zhi (2010-03-18)
Jun Bu, Xian-yi Zhuo
RESUMEN
To study the relation between human blood estazolam concentration and neurobehavioral function. The neurobehavioral ability of 10 volunteers were measured with computer-administered neurobehavioral evaluation system-chinese3 (NES-C3) and SMART EquiTest system. The neurobehavioral ability and balance function declined 1 h later after dosing estazolam. The neurobehavioral ability index and balance function declined to the lowest level 3 h later after dosing estazolam. The neurobehavioral ability recovered partly 6 h later after dosing estazolam, and neurobehavioral ability recovered completely 10 h later. Driving ability was impaired when estazolam concentration in blood is 20 ng/mL, and the neurobehavioral ability declined when estazolam concentration is 40 ng/mL in blood. The influence to human in absorption process is greater than the metabolic process with the same estazolam concentration.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Estazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Estazolam