MilliporeSigma
  • Nickel-catalyzed reductive cleavage of aryl-oxygen bonds in alkoxy- and pivaloxyarenes using hydrosilanes as a mild reducing agent.
Nickel-catalyzed reductive cleavage of aryl-oxygen bonds in alkoxy- and pivaloxyarenes using hydrosilanes as a mild reducing agent.
Chemical communications (Cambridge, England) (2011-01-26)
Mamoru Tobisu, Ken Yamakawa, Toshiaki Shimasaki, Naoto Chatani
RESUMEN
A nickel-catalyzed reductive deoxygenation of aryl alkyl ethers and aryl pivalates has been developed. Hydrosilanes serve as a mild reducing agent. The present protocol allows the use of a pivalate group as a robust and traceless steering group in arene functionalization reactions.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Pivalic acid, 99%