MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Activation of presynaptic GABAB receptors modulates GABAergic and glutamatergic inputs to the medial geniculate body.
Hearing research (2011-06-15)
Bin Luo, Hai-Tao Wang, Yan-Yan Su, Shu-Hui Wu, Lin Chen
RESUMEN
The medial geniculate body (MGB) receives ascending inputs from the inferior colliculus and descending inputs from the auditory cortex. In the present study, we intended to determine whether activation of presynaptic GABA(B) receptors modulates GABAergic and glutamatergic inputs to the MGB with whole-cell patch-clamp recordings in brain slices of the rat. To evoke a synaptic response, we electrically stimulated the ascending and descending inputs to MGB neurons with bipolar electrodes placed on the brachium of the inferior colliculus and the superior thalamic radiation. To isolate presynaptic mechanisms, we blocked the effects of postsynaptic GABA(B) receptors by filling recording electrodes with the internal solution containing cesium and QX-314. The activation of presynaptic GABA(B) receptors by exogenous agonist was shown to modulate synaptic inputs to the MGB as demonstrated by that (1) baclofen, a GABA(B) receptor agonist, reversibly suppressed both inhibitory postsynaptic currents (IPSCs) and excitatory postsynaptic currents (EPSCs) and this suppressive effect could be blocked by CGP35348, a GABA(B) receptor antagonist, (2) baclofen significantly increased the ratio of IPSCs or EPSCs elicited by paired-pulse stimulation, and (3) baclofen depressed EPSCs and IPSCs in response to repetitive stimulation. The activation of presynaptic GABA(B) receptors by endogenously released GABA was shown to modulate the synaptic transmission as demonstrated by that CGP55845, another GABA(B) receptor antagonist, increased the ratio of IPSCs to paired-pulse stimulation in young (P8-10) rats, although not in juvenile (P15-18) rats. Our study provides electrophysiological evidence for the presence of functional presynaptic GABA(B) receptors in the MGB and suggests an age-dependent role of these receptors in the synaptic transmission in this central auditory region.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
CGP 35348 hydrate, ≥97% (NMR), solid