MilliporeSigma
Nonenzymatic amperometric organic peroxide sensor based on nano-cobalt phthalocyanine loaded functionalized graphene film.
Analytica chimica acta (2011-12-20)
Lin Cui, Lijian Chen, Minrong Xu, Haichao Su, Shiyun Ai
RESUMEN
An enzyme-free amperometric method was established for the electrochemical reduction tert-butyl hydroperoxide (TBHP) on the utilization of nano-cobalt phthalocyanine (CoPc) loaded functionalized graphene (FGR) and to create a highly responsive organic peroxide sensor. FGR was synthesized with a simple and fast method of electrolysis with potassium hexafluorophosphate (KPF(6)) solution as electrolyte under the static current of 0.2A. In the present work, FGR was dispersed in the solution of CoPc to fabricate chemical modified electrode to detect TBHP. The electro-reduction of TBHP can be catalyzed by FGR-CoPc, which has an excellent electrocatalytical activity due to the synergistic effect of the FGR with CoPc. The sensor can be applied to the quantification of TBHP with a linear range covering from 0.0260 to 4.81 mM, a high sensitivity of 13.64 A M(-1), and a low detection limit of 5 μM. This proposed sensor was designed as a simple, robust, and cheap analytical device for the determination of TBHP in body lotion.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cobalt(II) phthalocyanine, β-form, Dye content 97 %