MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Synthesis and evaluation of changes induced by solvent and substituent in electronic absorption spectra of some azo disperse dyes.
Synthesis and evaluation of changes induced by solvent and substituent in electronic absorption spectra of some azo disperse dyes.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-01-21)
Asadollah Mohammadi, Mohammad Reza Yazdanbakhsh, Lahya Farahnak
RESUMEN
Five azo disperse dyes were prepared by diazotizing 4'-aminoacetophenone and p-anisidine and coupling with varies N-alkylated aromatic amines. Characterization of the dyes was carried out by using UV-vis, FTIR and 1H NMR spectroscopic techniques. The electronic absorption spectra of dyes are determined at room temperature in fifteen solvents with different polarities. The solvent dependent maximum absorption band shifts, were investigated using dielectric constant (ɛ), refractive index (n) and Kamlet-Taft polarity parameters (hydrogen bond donating ability (α), hydrogen bond accepting ability (β) and dipolarity/polarizability polarity scale (π*)). Acceptable agreement was found between the maximum absorption band of dyes and solvent polarity parameters especially with π*. The effect of substituents of coupler and/or diazo component on the color of dyes was investigated. The effects of acid and base on the visible absorption maxima of the dyes are also reported.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
p-Anisidine, 99%
Sigma-Aldrich
4′-Aminoacetophenone, 99%