MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Direct synthesis of inverse hexagonally ordered diblock copolymer/polyoxometalate nanocomposite films.
Journal of the American Chemical Society (2012-07-05)
Thomas Lunkenbein, Marleen Kamperman, Zihui Li, Carina Bojer, Markus Drechsler, Stephan Förster, Ulrich Wiesner, Axel H E Müller, Josef Breu
RESUMEN
Nanostructured inverse hexagonal polyoxometalate composite films were cast directly from solution using poly(butadiene-block-2-(dimethylamino)ethyl methacrylate) (PB-b-PDMAEMA) diblock copolymers as structure directing agents for phosphomolybdic acid (H(3)[PMo(12)O(40)], H(3)PMo). H(3)PMo units are selectively incorporated into the PDMAEMA domains due to electrostatic interactions between protonated PDMAEMA and PMo(3-) anions. Long solvophilic PB chains stabilized the PDMAEMA/H(3)PMo aggregates in solution and reliably prevented macrophase separation. The choice of solvent is crucial. It appears that all three components, both blocks of the diblock copolymer as well as H(3)PMo, have to be soluble in the same solvent which turned out to be tetrahydrofuran, THF. Evaporation induced self-assembly resulted in highly ordered inverse hexagonal nanocomposite films as observed from transmission electron microscopy and small-angle X-ray scattering. This one-pot synthesis may represent a generally applicable strategy for integrating polyoxometalates into functional architectures and devices.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Phosphomolybdic acid solution
Sigma-Aldrich
Polybutadiene, average Mn ~5,000
Sigma-Aldrich
Phosphomolybdic acid solution, spray reagent, 10% in ethanol
Sigma-Aldrich
Polybutadiene, cis, average Mw 200,000-300,000
Sigma-Aldrich
Polybutadiene, predominantly 1,2-addition, approx. 90% 1,2-vinyl
Sigma-Aldrich
Phosphomolybdic acid solution, 20 wt. % in ethanol
Sigma-Aldrich
Phosphomolybdic acid solution, ready-to-use spray and plunge reagent
Sigma-Aldrich
Sodium phosphomolybdate hydrate, technical