MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Determination of cymoxanil in drinking water and soil using high-performance liquid chromatography.
Biomedical chromatography : BMC (2005-05-26)
Paola Fidente, Carmen Di Giovanni, Serenella Seccia, Patrizia Morrica
RESUMEN
We describe a method for determination of cymoxanil, 1-2-cyano-2-methoxy(iminoacetyl)-3-ethylurea, in drinking water and in soil, using reversed-phase HPLC with UV detection at 240 nm and a mobile phase of acetonitrile-water (30:70, v/v). Fortified water samples (1.0 L) were extracted with solid-phase extraction on Strata X. Soil samples (20 g) were extracted with acetone and the extracts were transferred onto Strata C18E. The recoveries of cymoxanil from water and soil samples were over 85% for each fortification level. The RDS were within the range 1.7-4.1% for water and 0.9-1.2% for soil samples. After optimization of the extraction and separation conditions, the method was validated.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Cymoxanil, PESTANAL®, analytical standard