MilliporeSigma
Single night postoperative prone posturing in idiopathic macular hole surgery.
European journal of ophthalmology (2011-09-20)
Arshed Malik, Ian Dooley, Usman Mahmood
RESUMEN
To evaluate the role of postoperative prone posturing for a single night in the outcome of trans pars plana vitrectomy (TPPV) with internal limiting membrane (ILM) peel and 20% perfluoroethane (C2F6) internal tamponade for idiopathic macular hole. This prospective trial enrolled 14 eyes in 14 consecutive patients with idiopathic macular hole. All eyes underwent TPPV with vision blue assisted ILM peeling with and without phacoemulsification and intraocular lens (IOL) for macular hole. Intraocular gas tamponade (20% C2F6) was used in all cases with postoperative face-down posturing overnight and without specific posturing afterwards. LogMAR visual acuity, appearance by slit-lamp biomicroscopy, and ocular coherence tomography (OCT) scans were compared preoperatively and postoperatively to assess outcome. Among 14 eyes recruited, all eyes were phakic; 50% of patients underwent concurrent phacoemulsification with IOL. The macular holes were categorized preoperatively by OCT appearance, 4 (28.57%) were stage 2, 7 (50%) were stage 3, and 3 (21.43%) were stage 4. Mean macular hole size was 0.35 disk diameters. Symptoms of macular hole had been present for an average of 6.5 months. All holes (100%) were closed 3 and 6 months postoperatively. Mean visual acuity (logMAR) was improved to 0.61 at 3 months and was stable at 6 months after the surgery. None of the eyes had worse vision postoperatively. Vitrectomy with ILM peeling and 20% C2F6 gas with a brief postoperative 1 night prone posturing regimen is a reasonable approach to achieve anatomic closure in idiopathic macular hole. Concurrent cataract extraction did not alter outcomes and was not associated with any additional complications.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Hexafluoroethane, ≥98%