MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Labdane diterpenoids and lignans from Calocedrus macrolepis.
Fitoterapia (2013-01-29)
Xing-De Wu, Shuang-Yan Wang, Lei Wang, Juan He, Gen-Tao Li, Lin-Fen Ding, Xun Gong, Liao-Bin Dong, Liu-Dong Song, Yan Li, Qin-Shi Zhao
RESUMEN
Three new labdane diterpenoids, calomacrins A-C (1-3), and a new diaryl butyrolactone-type lignan, calomacrol A (8), as well as four known labdane diterpenoids and six known lignans, were isolated from the twigs and leaves of Calocedrus macrolepis. Structures of the new compounds were elucidated on the basis of their spectroscopic methods, including 1D and 2D NMR techniques. Compounds 3-14 were evaluated for cytotoxicity against HL-60, SMMC-7721, A-549, MCF-7, and SW480 cell lines.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
γ-Butirolactona, ReagentPlus®, ≥99%
Sigma-Aldrich
γ-Butirolactona, ≥98%, FCC, FG
Sigma-Aldrich
γ-Butirolactona, SAJ first grade, ≥99.5%