MilliporeSigma
  • Application of the bivariate spectrophotometric method for the determination of metronidazole, furazolidone and di-iodohydroxyquinoline in pharmaceutical formulations.
Application of the bivariate spectrophotometric method for the determination of metronidazole, furazolidone and di-iodohydroxyquinoline in pharmaceutical formulations.
Journal of pharmaceutical and biomedical analysis (1997-12-31)
P L López-de-Alba, K Wróbel, L López-Martínez, K Wróbel, M L Yepez-Murrieta, J Amador-Hernández
RESUMEN
The bivariate calibration algorithm was applied to the spectrophotometric determination of metronidazole, furazolidone and di-iodohydroxyquinoline in pharmaceutical dosage forms. The results obtained were compared with the results of derivative spectrophotometry. The statistical evaluation of method bias was carried out, and it was shown that the proposed procedure may be competitive with commonly used first-derivative spectrophotometry. The advantage of the bivariate calibration is its simplicity, and the fact that there is no need to use the derivatization procedures.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
5,7-Diiodo-8-hydroxyquinoline, 95%