MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Transdermal delivery of levosimendan.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2000-10-18)
R Valjakka-Koskela, J Hirvonen, J Mönkkönen, J Kiesvaara, S Antila, L Lehtonen, A Urtti
RESUMEN
The aim of this study was to determine if transdermal penetration of levosimendan, a novel positive inotropic drug, could be enhanced and controlled by formulation modifications. Penetration of levosimendan across human epidermis in vitro was determined using abdominal excised skin and diffusion cells. Predicted steady-state plasma concentrations of levosimendan were estimated using permeabilities and pharmacokinetic parameters of levosimendan. For penetration enhancement we used different pH values, co-solvents, cyclodextrins, surfactants, penetration enhancers, liposomes, and iontophoresis. Sodium lauryl sulfate, ethanol, oleic acid, and soya phosphatidylcholine or their combinations clearly increased levosimendan permeation across the skin in vitro. Iontophoresis was also an efficient method to increase transdermal permeation of levosimendan. A hydrophilic co-solvent/penetration enhancer is needed to achieve better permeability of levosimendan across the skin. In conclusion, transdermal delivery of levosimendan can be significantly increased by formulation modification. Based on kinetic calculations, therapeutic plasma concentrations may be achievable transdermally.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido oleico, technical grade, 90%
Sigma-Aldrich
Ácido oleico, ≥99% (GC)
Sigma-Aldrich
Ácido oleico, BioReagent, suitable for cell culture
Sigma-Aldrich
Ácido oleico, natural, FCC
Sigma-Aldrich
Ácido oleico, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Ácido oleico, analytical standard
Sigma-Aldrich
Ácido oleico, SAJ first grade, ≥70.0%
Supelco
Ácido oleico, Selectophore, ≥99%