MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Amino acid levels and ratios in serum and cerebrospinal fluid of patients with optic neuropathy in Cuba.
Nutritional neuroscience (2002-02-15)
A Gonzalez-Quevedo, F Obregon, R Fernandez, R Santiesteban, C Serrano, L Lima
RESUMEN
Twenty-one amino acids were determined in serum and cerebrospinal fluid of 12 patients with endemic, and in the cerebrospinal fluid of 22 patients with epidemic optic neuropathy. For the endemic patients, there was a decrease in aspartate and taurine in the serum with respect to controls. The ratios aspartate/taurine and taurine/valine were decreased, and glutamate/taurine was increased in the serum. Some of the altered amino acid ratios indicate preponderance of excitatory to inhibitory molecules. The ratio with valine corresponded to the decrease in taurine and the maintenance of valine concentration, an amino acid related to anthropometric parameters. A typical malnutrition pattern was not observed, as the levels of essential amino acids were not significantly modified. In the cerebrospinal fluid there were increases in aspartate, glutamate and threonine, the first two probably indicating a neurodegenerative disorder or some type of metabolic alteration, primary or secondary to the disease. The increase in threonine could be related to lipid metabolism, but it is not clear at present. A wide variety of amino acid ratios were increased in the cerebrospinal fluid of patients with endemic optic neuropathy, mainly pointing to an excitatory condition and some metabolic alterations. In the cerebrospinal fluid of patients with epidemic optic neuropathy there was an increase in aspartate and glutamate, and increase in glutamate/taurine, glutamate/glycine, and gamma-aminobutyric acid/glycine ratios. Interesting differences were also observed between patients from different periods of time, but with the same clinical features, and the modifications of amino acid concentrations in the cerebrospinal fluid, such as glutamine, threonine and tryptophan. The present results indicate a disorder in the metabolism of amino acids, support a specific deficit, especially for taurine, an imbalance between excitatory and inhibitory amino acids, and a possible relation to viral infections.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Taurine, ≥98%, FG
SAFC
Taurine