MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Lipid peroxidation and vitamin E in human coronary atherosclerotic lesions.
Clinica chimica acta; international journal of clinical chemistry (2003-03-15)
Wojciech Jacheć, Andrzej Tomasik, Wojciech Ceglarek, Stanisław Woś, Jan Wodniecki, Celina Wojciechowska, Bronisława Skrzep-Poloczek, Piotr Walichiewicz, Kazimierz Widenka
RESUMEN
We aimed to assess the oxidant/antioxidant status within the ex vivo human coronary endarterectomy samples. To achieve this, we measured products of lipid oxidation (malondialdehyde, 7-ketocholesterol), lipids (cholesterol, cholesteryl esters) and vitamin E in endarterectomy samples. Content of malondialdehyde in the plaque ranged from 0.23 to 37.36 microg/g. Unesterified cholesterol content ranged from 0.30 to 1.94 mg/g. It was 9.04+/-4.32% of total cholesterol. Total cholesterol content ranged from 1.73 to 23.69 mg/g. Cholesteryl palmitate content ranged from 0.57 to 19.10 mg/g, which is 11.43-60.86% of the total esters (mean+/-SD 40.27+/-18.42%). Cholesteryl oleate content ranged from 0.24 to 5.76 mg/g, being 9.97-21.81% of total esters (mean+/-SD 14.35+/-4.51%). Cholesteryl linoleate content ranged from 1.05 to 8.21 mg/g, being 17.84-45.15% of total esters (mean+/-SD 30.78+/-11.69%). Cholesteryl arachidonate content ranged from 0.51 to 4.20 mg/g, which is 7.56-22.87% of total esters (mean+/-SD 14.60+/-5.60%). The cholesteryl linoleate/cholesteryl oleate ratio (CL/CO) ranged from 1.01 to 4.33. Content of 7-ketocholesterol in the plaque ranged from 0.0 to 577.5 ng/g of wet weight. The 7-ketocholesterol/total cholesterol ratio was 0.003+/-0.003% (range from 0.0% to 0.008%). The 7-ketocholesterol/unesterified cholesterol ratio was 0.024+/-0.023% (range from 0.0% to 0.066%). The plaque content of vitamin E ranged from 0.0 to 40.9 microg/g of wet weight. The present study, comprising measurements of lipids, products of lipid peroxidation and vitamin E in 12 human coronary endarterectomy samples, lends the evidence for ongoing lipid peroxidation within an atherosclerotic lesion.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cholesteryl palmitate, ≥98% (HPLC; detection at 205 nm)